Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bulb pipettes, Class AS/Class A

Bulb pipettes, Class AS/Class A

Bulb pipettes, Class AS/Class A
Certificates
Bulb pipettes, Class AS/Class A
Bulb pipettes, Class AS/Class A

2 Options Available Below

Product Details & Documents

Bulb pipettes, conformity certified.

  • Fused amber graduation
  • Class AS within the official tolerance
Product Family Options

One mark

Product Information

  • Capacity
    Tolerance
    Length
    Colour code
    Suppl. to DIN
    Availability
    Price
  • HECH42556004
    Class AS, with ground-in plunger
    Class AS, with ground-in plunger
    0,5 ml
    ±0,005 ml
    300 mm
    2 × black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,60 €
    4,60 €
    1 * 1 ST
       4,60 €

Two marks

Product Information

  • Capacity
    Tolerance
    Length
    Colour code
    Suppl. to DIN
    Availability
    Price
  • HECH42557024
    Class A, with ground-in plunger
    Class A, with ground-in plunger
    3 ml
    ±0,010 ml
    350 mm
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,60 €
    4,60 €
    1 * 1 ST
       4,60 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.