ContactUsLinkComponent
Counting chambers, Jessen
Counting chambers, Jessen
Catalog # HECH40450702
undefined
Counting chambers, Jessen
Catalog # HECH40450702
Specifications
  • Depth of chamber:
    0,4 mm
  • Type:
    Jessen
  • Description:
    Counting chamber (CE)
  • Seq:
    001
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
These double ruling chambers are used for blood tests, urine analysis, and counting cells in cerebrospinal fluid (CSF).