Counting chamber, Bürker-Türk
Catalog # HECH40444702
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Depth of chamber:0,1 mm
- Type:Bürker-Türk
- Description:Cell counter
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Double ruling chambers for blood tests and urine analysis
- Groups of squares in the corners divided by double lines
- Overall size and the central square correspond to the Neubauer ruling