Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Connexion
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lids
Microplate Lids
Lids for 96-well microtest plates
Lids for 96-well microtest plates
Lids for 96-well microtest plates
Catalog # SARS82.1584
Fournisseur:
SARSTEDT
Expand image
Lids for 96-well microtest plates
Catalog # SARS82.1584
Fournisseur:
SARSTEDT
Spécifications
Colour:
Clear
Description:
Lids for micro test plates, PS
Pk:
100
Spécifications
À propos de cet article
Product Details
Product Details