Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dissecting needles

Dissecting needles

Dissecting needle, handle length 100 mm, Straight, 140 mm
Catalog # HECH42712010
Dissecting needles
Catalog # HECH42712010
Dissecting needles
Dissecting needles

Dissecting needle, handle length 100 mm, Straight, 140 mm

Price
2,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Dissecting needles made of chromium-nickel steel, stainless, with wooden handle.

Specifications
  • Form:
    Straight
  • Description:
    Dissecting needle, handle length 100 mm
  • Length:
    140 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.