HPLC columns, PLRP-S
Supplier: VARIAN
Certificates
About this item
The PLRP-S family of products consists of a range of pore sizes and particle sizes, all with identical chemistry and fundamental adsorptive characteristics. The particles are inherently hydrophobic so there is no bonded phase, alkyl ligand required for reversed-phase separations. This gives a highly reproducible material that is free from silanols and heavy metal ions. Within the extensive product range are columns suitable for analytical separations and preparative purifications.
- Contain durable and resilient particles that deliver reproducible results over longer lifetimes
- Thermally and chemically stable
- Used in bioscience, chemical, clinical research, energy, environmental, food and agriculture, material science and pharmaceutical industries
Buffer content: unlimited.
Organic modifier: 1 to 100%.
Maximum pressure: 5 to 8 μm: 210 bar; 3 μm: 300 bar.
Applications:
100 Å - Small molecules/synthetic biomolecules
300 Å - Recombinant peptides/proteins
1000 Å - Large proteins
4000 Å - DNA/high speed
Specifications
- Catalog No:
- 568-0818
- 568-0789
- 568-0815
- 568-0807
- 568-0833
- 568-0808
- 568-0835
- 568-0823
- 568-0836
- 568-0824
- 568-0834
- 568-0830
- 568-0831
- 568-0832
- 568-0814
- 568-0827
- 568-0828
- 568-0806
- 568-0829
- 568-0816
- 568-0817
- 568-0821
- 568-0813
- 568-0822
- 568-0825
- 568-0826
- 568-0837
- 568-0810
- 568-0811
- 568-0820
- 568-0812
- 568-0798
- 568-0799
- 568-0800
- 568-0801
- 568-0809
- 568-0838
- 568-0802
- 568-0803
- 568-0804
- 568-0805
- pH range:
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- Temperature limit:
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- 200 °C
- USP code:
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
Specifications
Frequently Bought Together