ContactUsLinkComponent
Cell culture plates with Cell+ surface
Specifications
  • No. of wells:
    24
  • Description:
    Cell culture plates, Cell+ growth surface
  • Environmentally Preferable:
  • Culture area:
    2 cm²
  • Pk:
    50
  • Well volume:
    3,6 ml
Specifications
About this item
PS, gamma sterilised, with lid.
  • Wells with raised rims
  • Stackable
  • Lid with condensation rings
  • Lid can only be positioned in one direction, avoids cross contamination
Cell+ TC plates for the growth of primary cultures and other fastidious cell lines.