Orders LinkContactUsLinkComponent
Splash heads, with sinter, Quickfit®
Splash heads, with sinter, Quickfit®
Supplier:   SciLabware
Splash heads, with sinter, Quickfit®
Supplier:  SciLabware
Certificates
Splash heads, with sinter, Quickfit®
Splash heads, with sinter, Quickfit®
3 Options Available Below
Product Details & Documents
PYREX® borosilicate glas
  • Prevents possible contamination of the condenser from 'bumping' solvents
  • Features a coarse porosity 1 sinter to reduce carry over
  • Splash heads feature a 29/32 socket
For use with rotary evaporators.
Product Family Options
Product Information
  • Socket size
    Cone size
    Availability
    Price
  • 29/32 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 901,00 €
    901,00 €
    1 * 5 ST
       901,00 €
  • 29/32 NS
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 901,00 €
    901,00 €
    1 * 5 ST
       901,00 €
  • 29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 181,00 €
    181,00 €
    1 * 1 ST
       181,00 €