Orders LinkContactUsLinkComponent
csm_dm2500-3_02_04692c24fe_200x200.jpg
Leica Compound Microscopes
Get insights into the smallest details and inspect and document results efficiently with Life Science  compound microscopes from Leica Microsystems. Each solution can be customized with brilliant, cool LED illumination, ergonomic accessories, sophisticated digital cameras and intuitive software to meet a broad range of applications.
Leica Compound Upright Microscopes | Avantor
Product selection
leica-dm300_100x100.jpg
Leica DM300
The Leica DM300 compound microscope is compact, Maintenance-free and easy to use to provide trouble free operation each day. A routine Life Sciences microscope solution.
csm_Leica_DM1000_LED_Life_8b73115a58_100x100.jpg
The Leica DM1000 LED combines optical excellence with ergonomic design and features long-life LED illumination that provides near daylight, bright illumination with constant color temperature and emits less heat...
Leica_DMi1_07_100x100.jpg
The Leica DMi1 inverted microscope supports your specific work routine in your cell culture lab. Its operation is so intuitive, its handling so comfortable that you can fully concentrate on your work. Choose the functions you need, and, if necessary...
DM-IL-LED_16_100x100.jpg
With high-performance optics, ergonomic design, and 5W LED illumination, the Leica DM IL LED is ideal for cell culture, micromanipulation, documentation of immunostained specimens, and routine live cell examinations.
csm_ThunderImager-Tissue_5a35b2581a_100x100.jpg
The THUNDER Imager Tissue allows real-time fluorescence imaging of 3D tissue sections typically used in neuroscience and histology research. Acquire rich, detailed images of thick tissues free of haze from out-of-focus blur.
THUNDER-Imager-3D-Cell-Culture_100x100.jpg
THUNDER Imagers provide you with a solution for advanced 3D cell culture assays, whether you want to study stem cells, spheroids, or organoids.