Orders LinkContactUsLinkComponent
brady3-hero.jpg
Brady - Simplify your laboratory identification
Simplify Your Laboratory Identification With Brady Solutions
Brady’s laboratory identification products are designed to serve the world’s biotechnology, agricultural, environmental and forensic researchers. Whether you work in pathology, histology, chromatography or other areas of science, Brady labelling solutions can help you maintain Good Laboratory Practice.
Brady - Simplify your laboratory identification | Avantor
Brady Sample Identification Solutions
  • Match existing vial, slide and well plate size
  • Remains legible throughout processing and storage
  • Endures freezer storage, liquid nitrogen, autoclaves, hot water baths and solvents, including xylene, DMSO and ethanol
  • Allows barcoding for extensive information storage on even the tiniest sample
  • Integrates seamlessly with your Laboratory Information Management System (LIMS)
brady-rfid-540.jpg
Bring your workplace to the next level of efficiency and productivity with reliable RFID.
brady-new-product-540.jpg
Discover more reliable solutions to protect and identify people, products and places for mission critical applications in harsh environments.
brady-sample_identification-540.jpg
Printed laboratory labels engineered specifically to identify samples, help avoid sample loss, and a lot of frustration!
onsite-540.jpg
The ability to print your labels on-site enables you to quickly respond to any identification need, and can eliminate waiting on label deliveries!
sample-id-540.jpg
Brady`s printer applicators offer an automated solution to applying 'print on demand' labels!
brady-safety-540.jpg
Make your workplace compliant, safer and more efficient!
Brady HQ
brady-building-540.jpg
Brady Corporation is an international manufacturer and marketer of complete solutions that identify and protect people, products and places.
At Brady, commitment to performance drives our business decisions every day. We design our products and solutions to perform in ways that others simply don't. Our people perform in ways that regularly exceed expectations. And our focus and discipline as a company works toward solid, long-term performance for our customers and shareholders. In short, the essence of Brady, and subsequently our brand, is centerd on 'performance'.
About Brady