Orders LinkContactUsLinkComponent
AzureCielo955x590.jpg
Azure Cielo™ 3 and Cielo 6
The Azure Biosystems Cielo 3 and 6 real-time PCR Systems are designed to offer smart protocols, high quality data, robust durability and an intuitive user experience.
Azure Cielo™ 3 and Cielo 6 | Avantor
Our novel fiber optic detection system allows for enhanced sensitivity and speed. 3 or 6 standard channels let you analyze a variety of commercially available dyes, already in your workflow. Onboard software operated by a 10.2 touchscreen minimizes footprint and saves you bench space.
Easily connect to your Cielo via Wi-Fi, ethernet, USB or LAN. Get notified after each completed run and even remotely access protocols and files.