Orders LinkContactUsLinkComponent
Lauda products | Avantor
Lauda products
Circulating chillers.png
Range of circulation chillers
includes five compact models
providing cooling capacities from
0,25 to 1,2 kW for external
applications. The units are ideal
for use where heat needs to be
dissipated reliably and quickly.
Heating thermostats with transparent baths.png
ECO units with transparent
plastic baths provide visibility
where test samples need to be
observed. Flow rate switch for
flow rate control between internal
and external circulation.
Circulation thermostats.png
The LOOP product range
includes two thermo-electric
circulation thermostats for
external temperature control.
Refrigerant-free temperature
control with Peltier technology.