Orders LinkContactUsLinkComponent
Digital Intelligent Measurement for more Accuracy and Flexibility! | Avantor
Digital Intelligent Measurement for more Accuracy and Flexibility!
wtw_sentix_main.jpg
Initially pH measurement started with a reference electrode and a measuring cell. In time, the combined electrode took over, followed by the combined electrode with built-in temperature sensor. Today, we take another step into the future with the innovative Digital sensors for pH, redox, conductivity, dissolved oxygen and even turbidity.
Digital Intelligent sensors are:
  • More accurate as the signal is converted directly in the electrode
  • Intelligent as serial numbers and settings including calibration dates are transferred automatically to the meter for best data management ever
  • Easy to use as each unique sensor is recognised by the meter
and much more ....
undefined
VWR® Multiparameter Digital MD 8000
Advantages of a Digital measurement system
Watch this video to see how these innovations make your work easier, more accurate and give you flexibility for future projects!
Watch this video
Product selection
phenomenal_md_8000L.jpg
One channel bench meter
6 parameters: pH/temperature, redox, conductivity, TDS, Salinity, Dissolved oxygen and turbidity
Memory 5000 Datasets GLP
phenomenal_md_8000h.jpg
One channel portable meter
6 parameters: pH/temperature, redox, conductivity, TDS, Salinity, Dissolved oxygen and turbidity
Memory 5000 Datasets GLP
Product selection
wtw_ids_lab_meters.jpg
1 to 3 channel bench meter
6 parameters: pH/temperature, redox, conductivity, TDS, Salinity, Dissolved oxygen and turbidity.
Memory 5000 Datasets GLP
wtw_ids_portable_meters.jpg
1 to 3 channel portable meter
6 parameters: pH/temperature, redox, conductivity, TDS, Salinity, Dissolved oxygen and turbidity
Memory 5000 Datasets GLP
HACH® Single- & Multi-parameter HQd systems
HQD_meter.jpg
HQD meter, Have a look at Hach's HQd IntelliCAL durable water testing probes.
HQD_mono_and_multi_parameter.jpg
1- to 2-channel meter: pH/temperature, redox, conductivity, TDS, salinity, dissolved oxygen and ISE. 500 data set GLP memory.
HQD_portable_meters.jpg
1- to 2-channel meter: pH/temperature, redox, conductivity, TDS, salinity, dissolved oxygen and ISE. 500 data set GLP memory.