Orders LinkContactUsLinkComponent
Karl-Fischer.jpg
Karl Fischer Titration
Avantor® offers a choice of compact volumetric or coulometric Karl Fisher titrators and related reagents for water determination.
  • Volumetric Karl Fischer titrators are for medium to large volumes of water (mg range) combining fast and precise water content determination with easy operation
  • Coulometric Karl Fischer titrators are for small water volumes (µg range) combine fast and precise water content determination with very simple operation
  • Karl Fischer reagents
Karl Fischer Titration | Avantor
Product selection
thumb_kf_titrator_1_100x90.jpg
Karl Fisher Titrators
Routine coulometric or volumetric Karl Fisher titrators combine fast and precise water content determination with very simple operation.
thumb_kf_reagents.png
Reagents for the determination of water by Karl Fischer titration of a wide range of substrates and a variety of methodologies