Orders LinkContactUsLinkComponent
29969_density-measurement-1920x760.jpg
Density measurement
Densimeters can be used in many applications, including quality control of incoming goods and quality assurance of intermediate or final products. The choice of technique for sample analysis depends on the sample type, the required accuracy and ease of use. The Mettler density guide provides information on the preferred methods for handling liquids. For solid and liquid samples, balances with built-in density functions or under-hook weighing functions can be used to manually determine density.
Density measurement | Avantor
Featured products
digital-hydrometer-540x390.jpg
Handheld digital hydrometers, or digital density meters, are used to determine the density of liquids quickly and automatically.
density-kits-for-vwr-balances-540x390.jpg
Complete kits that, together with the appropriate balance, let users calculate the density of liquids or non-absorbent solids.
density-standards-540x390.jpg
Density standards can be used as calibration standards for density measurements performed with pycnometric, vibrational or hydrometer-based techniques.
hydrometers-540x390.jpg
Hydrometers and pycnometers measure the specific gravity or relative density of liquids or solids.
BELL43-xx-DSG-Density-Meter-540x390.jpg
DSG Series density meters are fitted with Peltier temperature control for measuring the density, specific gravity, Brix and other parameters of liquid solutions.
Have questions about density measurement products?