Orders LinkContactUsLinkComponent
mastersense-bt-biopharma-pump-1920x760.jpg
Masterflex® B/T® MasterSense™ BioPharma Process Pump
Reimage bioprocessing
NEW integrated sensor technology, intuitive touchscreen and MasterflexLive® remote monitoring for the biopharma industry of tomorrow
Shop now
Masterflex® B/T® MasterSense™ BioPharma Process Pump | Avantor
MasterSense™ monitors your media so you don't have to
With the extra time, a process engineer could:
MasterSense_monitors_your_media_so_you_dont_have_t.png
Advanced Connectivity and Control Options
Remote, secure, and in compliance
bt-connectivity-img-diagram.png
Masterflex B/T MasterSense Complete Pump, up to 42LPM, IP66 Item #   MFLX77112-10
IP 66UL listedcandian UL listed2 year warranty
What's Included
Rapid-Load® pump head, 3 ft (1 m) of Tygon® LFL B/T 91 tubing 06430-91; and a 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. Unit is shipped with country specific plug/cord set; please specify ultimate destination when ordering.
Performance Specifications
Flow capacity0.65 to 42 LPM (0.17 to 11.1 GPM)
rpm11 to 321
Number of heads accepted1
Maximum torqueStarting: 1130 N•cm (1600 oz-in)
Running: 1017 N•cm (1440 oz-in)
ReversibleYes
External control - input0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10 V, 10 to 0 V, 0 to 5 V, 5 to 0 V; Start/Stop, Direction (CW/CCW), Prime via contact closure
External control - output0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10 V, 10 to 0 V, 0 to 5 V, 5 to 0 V; Tach output (open collector), Motor running, Open head, Local/Remote, General alarm
Network communicationEtherNet/IP, Profibus, MasterflexLive
Electrical Specifications
Voltage90 to 260, universal voltage autoselecting
Current5.0 A at 115 V; 3.1 A at 230 V
Fuse rating6 A / 250 V
Motor typeContinuous duty, TENV, BLDC
Motor size370 W (0.5 hp)
Display5” capacitive TFT/RGB touch screen with in-plane switching (wide viewing angle); 800 x 480 pixels
Motor/speed control typePWM with microprocessor
Speed regulation (repeatability)±0.1% 
Soft start/Electronic brakeYes/Yes
Physical specifications
Housing materials316 stainless steel housing, stainless steel chassis, stainless steel bezel
Pump head materialsPowder-coat carbon steel and cast aluminum, stainless steel, polycarbonate, Nylatron® rollers
IP ratingIP66
Agency listingsUL, cUL, CE, DLS
Operating temperature0 to 40°C (32 to 104°F)
Storage temperature-25 to 65°C (-13 to 149°F)
Noise level<75 dBA @ 1 m (39")
Dimensions (L x W x H)62.2 x 40.1 x 47.8 cm (24.5" x 15.8" x 18.8")
Shipping weight54.5 kg (120 lb)
Need on-demand pumping and portability?
WF274838_180_n.png
Designed for easy and secure operation of Masterflex pump systems, the new Masterflex Bioprocess Pump Cart is ideal for efficient operation in flexible manufacturing.
Learn more
The Perfect Pump for High-Volume Applications
Avantor_IP66WASHDOWN_GradientStroke-100.jpg
IP66 rated- built for sanitary, washdown environments including clean rooms, food, and dairy processing.
Avantor_Non-contaminating_Gradient-100.jpg
Non-contaminating, low shear, and noninvasive makes this pump perfect for Biopharma and healthcare.
Avantor_Flowrate_GradientStroke-100.jpg
Flows up to 42 LPM to ensure precise flow rates making this pump ideal for large-scale production.
Avantor_Locked_gradient-100.jpg
Three-tier access and the ability to save and recall programs making your fluidic process secure and accurate.
  • MasterflexLive is our secure, cloud-enabled platform for real-time remote monitoring of all Masterflex pump parameters
  • Monitor and adjust critical processes running 24/7 without the need for onsite personnel
  • Push notifications alert you to changes in operating conditions and error messages
Learn more about MasterflexLive
Flexible Connectivity
  • Your choice of connectivity options—MasterflexLive, EtherNet/IP, or Profibus
  • Three-tier access to pump functionality—use this option to protect your process by controlling who can operate the pump and at what level
  • Over-the-air firmware updates—automatically allows you to take full advantage of future enhancements
  • Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive
Intuitive Operation
bt-touchscreen-closeup-wet.jpg
Simple touch screen interface, even while wearing gloves
  • Easy to set up and program
  • 5" capacitive TFT/RGB touch screen with a clear, wide viewing angle
  • Adjust parameters with a simple touch—even with gloves on
  • Operational modes include continuous run, volume dispense, and time dispense
  • Batch control and interval/off time in Volume and Time modes
  • Easily save and recall up to 50 commonly used protocols, each with a unique name
  • Know when you hit your mark—graphical indicator lets you know when loaded tubing is optimally calibrated
  • IP66 rated for washdown
image_150x249.jpg
Continuous Mode
mastersense-bt-biopharma-time-sm.png
Time Mode
mastersense-bt-biopharma-volume-sm.png
Volume Mode
mastersense-bt-biopharma-ethernet-sm.png
Remote Mode
mastersense-bt-biopharma-tubcal-sm.png
Tube Calibration
mastersense-ls-tiereduser-sm.png
User Access
Connected, efficient and durable
MasterSense_BioPharma_Process_Pump.png
  1. Built for high-flow, continuous duty, perfect for sanitary environments — 316 polished stainless steel housing
    IP66 rated for washdown
  2. Integrated sensors monitor fluid pressure and alert before there is a potential for process failure
  3. Intuitive digital touchscreen with optimized menu navigation to store and recall standard protocols ensuring continuity - even with wet gloves on!
  4. Over-the-air firmware updates ensure optimization of your pump system without moving the system while MasterflexLive® cloud storage system securely stores, monitors and generates reports for all Masterflex pump systems
  5. Variable flow: 0.65 – 42 LPM
  6. Qualitative tube calibration ensures tubing is calibrated precisely – choose PerfectPosition® pump tubiung sizes B/T 87 and B/T 91
    SHOP B/T PUMP TUBING