Orders LinkContactUsLinkComponent
VWRC84209-955x590.jpg
Turbidity
Turbidity — the relative clarity of water — is a key indicator of quality for drinking water, wastewater, industrial water and more, making turbidity measurement an essential component of drinking water production.
Turbidity | Avantor
Public health and safety organizations worldwide recognize the importance of measuring turbidity. For example, a sudden change in turbidity could indicate biological, organic or inorganic pollution or signal a problem in the water treatment process.
Interference and errors can reduce the accuracy of any measuring instrument, so it is important to calibrate instruments to ensure that they are providing the most accurate information possible.
The documents listed below can help you achieve accurate readings when measuring turbidity.
Literature
Turbiditys_Role_In_EPA_Compliance___Hach_VWR-540x390.jpg
Turbidity measurement is important for compliance with U.S. Environmental Protection Agency regulations for drinking water.
Learn more.
Hach_Drinking_Water_Turbidity_Monitoring__7_Key_Considerations-540x390.jpg
Turbidity, as a measure of cloudiness or haze in water, has many useful applications. 
Learn more.
Hach_-_in_pursuit-of-perfect-glass-of-water_VWR-540x390.jpg
No isolated factor can ensure a perfect glass of water. The secret lies in
balancing multiple interrelated factors.
Learn more.
VWR_HACH_Turbidity_Science_540x390.jpg
An important water quality indicator for almost any use is the presence of dispersed, suspended solids. 
Learn more.
VWR_HACH_Turbidity_Science_540x390.jpg
All scientific work depends on precise measurement to assure meaningful results. 
Learn more.
LPV44299-tu52-540x390.jpg
The most precise and advanced turbidimeter on the market.
  • Patented 360°×90° optics
  • The most sensitive and fastest model available
  • The benchmark for clean water
Product details
AT-9681800_Turb-Lab-TL2300-Door-Opn-RZ-540x390.jpg
Get trusted measurements for a wide range of turbidity applications. A turbidimeter that simplifies testing in the most demanding wastewater and industrial applications.
  • Easy-to-use, full-color touchscreen
  • Easy data management and export capabilities
  • Watch product video
Delivered with sample cells, standards and accessories.
Product details
2100QIS-01_Turb-Lab-2100Qis-RZ-540x390.jpg
Ideal for use in the field or around the plant.
  • Easy to transport
  • Collects multiple measurements from various collection points in your treatment process
  • Intuitive user interface: easy to measure, calibrate and verify
  • Easy data storage and transfer 
Product details
tb210_vwr-540x390.jpg
For fast, precise and easy on-site testing.
  • From drinking water to wastewater
  • Can be used for colored and colorless liquids
  • Conforms to EN ISO 7027
Product details
31775499-540x390.jpg
Get optical sensitivity, reliable findings, ease of use and lab AQA with LIMS connection in this small but powerful unit that features:
  • Precise readings thanks to proven WTW optics
  • Intelligent reproducibility and plausibility check (IRPC)
  • Determination in drinking water according to DIN EN/U.S. EPA
Product details
AR-WTW-600700-Multi_VisoTurb_900P_Armatur-A925-K-540x390.jpg
VisoTurb® 900-P: digital IDS turbidity measurement with a practical turbidity sensor
  • Convenient turbidity sensor that measures turbidity in surface waters and during treatment processes
  • Simple 2- or 3-point calibration
  • Suitable for use with WTW MultiLine® IDS handheld meters for on-site measurements 
Product details
undefined
Our full range of turbidity meters
Need more info about our turbidity products?