Orders LinkContactUsLinkComponent
SustainableSolutions-955x980.jpg
Environmentally preferable safety and workwear
Without compromising on safety or welfare, there are now PPE and workwear options that are either manufactured in a more sustainable way, that use up less raw materials when produced and packaged, or that can be disposed of or recycled at end of use. Read more below.
Environmentally preferable safety and workwear | Avantor
Product selection
3M--Aura.jpg
3M™ Aura™ Particulate Respirators 9312+, 9322+ and 9332+ now contain at least 25% recycled content.
NPR15_9.jpg
Nitrile foam coating for excellent grip in wet and dry conditions. 15-gauge liner made from sustainable bamboo.
EcoSafety_8.jpg
Recycled knitted upper, made from recycled PET bottles. Recyclable green-thinking packaging.
HiVis_7.jpg
Hi-vis vests, jackets, trousers, fleeces, sweatshirts, polo shirts, t-shirts, made from 100% recycled polyester fabric.
PUREZERO-_5.jpg
Manufactured using 92% renewable energy. Dispenser boxes and outer cases are made using recycled materials.
MicroFlex31103_5.jpg
Bio based PLA (sugar based) compostable gloves with EN 13432 certification, made in Portugal. Achieve 90%+ biodegradation within 180 days in industrial composting facilities.
HyFlex11842_4.jpg
Multi-purpose gloves made from 90% recycled nylon/spandex liner. Carbon footprint reduction of 75 g CO₂ eq/pair. The plastic-free inner packaging contains 53% fewer materials.
V-Neck_140X140.jpg
Made from 100% recycled, high quality mono fiber polyesters, can be completely recycled. The recycled material can be made into new garments. Collection and recycling service is included in the cost of the garment.
Portwest_Biztex.jpg
Type 5/6 coveralls that can be washed 5 times.
cederroth_first_aid.jpg
The green-coloured plastic in Cederroth Stations and Dispensers are made of >85% recycled plastics.
green_line_shoe.jpg
The first collection of safety footwear to be given the EU Ecolabel.
25007479.jpg
The production process for these gloves has been redesigned to save gas, electricity and CO₂ emissions, so you can reduce your eco-footprint while still wearing a glove that is skin friendly, and provides superior barrier protection.
showa_biodgradable_gloves.jpg
Featuring SHOWA’s revolutionary Eco Best Technology®, these gloves are engineered for accelerated biodegradation in biologically active landfills.
8461402.jpg
These liners wick away perspiration, making gloves more comfortable for longer, so can be a way to reduce usage.
uvex_bamboo.png
Made from 45%; bamboo viscose and recycled polyamide. With reduced packaging and a CO2 neutral production rating.
glove-liners-uvex_200.jpg
Manufactured using Bamboo TwinFlex® technology which ensures maximum cut resistance, exceptional abrasion resistance and outstanding wearing comfort.
guard_eyewear_140.jpg
Modular eyewear range made using biobased and recycled materials, with improved packaging and the energy used in the production process is from biomass sources.
35942939.jpg
The nylon frame is manufactured from recycled fishing nets, with more environmentally friendly production, transportation and no plastic packaging.
18711406.jpg
Elten works with machines that are designed for energy and material-efficiency. So less waste is generated in the production process. Made in Germany, so less transport.
portwest_vegan_footwear.jpg
No animal products or animal by-products are used in the production of these footwear lines.
safety_footwear_140.jpg
Made from recycled materials with recycled packaging. Manufactured in a European facility using renewable energy and reduced waste.
25389632.jpg
Optional Eco packaging (bulk packed option) reduces waste and energy consumption.
36607433-140x140.jpg
A comprehensive range of safety helmets with compatible free recycling sheme (you only pay for the return shipping).
uvex_palanet.png
Shell is made from 30% bioplastic, headband is 50% recycled. Reduced and recycled packaging.