Orders LinkContactUsLinkComponent
battery-production.jpg
Battery Materials Production
Avantor® sets advanced battery science in motion to create a better world.
Battery Materials Production | Avantor
Critical Raw Materials, Chemicals, Equipment and Instruments for Battery Materials Production
Current Li-ion batteries consist of largely four main components: cathode, anode, electrolyte, and separator.
  • The cathode is a thin aluminum foil coated with a compound made up of active agent, solvent, super conductive additive and binder. Cathode active materials are typically composed of lithium and metal
  • The anode is typically a thin copper foil coated with a compound made up of active agent, solvent, super conductive additive and binder. Anode active material is graphite (synthetically-produced or mined)
  • The electrolyte is composed of salts, solvents and additives
  • Typical separators are currently made of synthetic resin such as polyethylene (PE) and polypropylene (PP)
Avantor will support you by offering critical raw materials, chemicals, equipment, instruments as well as a broad range of products for R&D, QA/QC, Testing, controlled environment, personal protective equipment, safety and facility management.
undefined
Contact your local sales representative for more information or fill in our contact form.
Featured Products: Personal Protective Equipment and Safety
Working-Clothes-495.jpg
Working clothes, disposable coverall, chemical protective coveralls.
Harness-495.jpg
A wide selection of harnesses.
gloves-495x400.jpg
Working clothes, disposable coverall, chemical protective coveralls.
GasDetector-495.jpg
Multi-gas detectors and oxygen detectors.
Featured Products: Chemicals, Raw Materials and Facility Management
LiPF6x495.jpg
The most common electrolyte in lithium batteries is a lithium salt solution such as lithium hexafluorophosphate (LiPF6).
Chemica--Storagex495.jpg
Our chemical storage solutions provide safe way to store corrosive or flammable materials, protecting personnel and facilities...
spill-kitx495.jpg
Our wide range of stationary systems and handheld wiping units can help you keep your laboratory space clean of spilled chemical solution or run-off. Look at the product Avantor offers for general-purpose spill control or specific applications.
Avantor offers a broad range of cleanroom products to protect the product and process.
Avantor offers a broad range of Personal Protective Equipment (PPE) to protect the operator when researching, manufacturing or recycling.
Avantor offers a broad range of products to keep the facility clean, safe and ordered.
Avantor offers a broad range of chemicals for R&D and production of battery active materials.
Avantor distributes a broad range of process equipment and instrumentation from top manufacturers.
Avantor carries a broad range of products to meet the specialized needs of R&D, QA/QC & Testing laboratories.