Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Equipment-Covers | Avantor
Equipment Covers
297.png
Tyvek® Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size., untreated, flexible Tyvek® material specifically designed for pharmaceutical use in steam sterilization applications.
327.png
Each closure incorporates a Tyvek® drawstring closure to create the tightest possible seal around all openings.
300 (2).jpg.png
Autoclavable up to 135 °C, BHD Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses.
321.png
Autoclavable up to 135 °C, each closure incorporates a Tyvek® drawstring closure to create the tightest possible seal around all openings.
Super Duty SMS Muffs.png
Designed for autoclave cycles (121 to 135 °C, Super Duty SMS Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses.
heavy duty muff.png
Designed for autoclave cycles (121 to 135 °C, Heavy Duty SMS Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses.
Tyvek skirted Muff1.png
The skirt extension on these Muffs provides a safe area to remove the it without inadvertently touching the critical surfaces with the fingers.
634.png
Leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
1088.png
Autoclavable up to 135 °C, leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
sd sms bowl cover2.png
Designed for autoclave cycles (121 to 135 °C) leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
HD sms bowl cover2.png
Designed for autoclave cycles (121 to 135 °C) leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
309.png
Keeps biopharmaceutical pharmaceutical production equipment clean and protected from potentially damaging UV light.
418.png
Helps minimize microbial and particulate contamination risks in pharmaceutical production operations.
495.png
GMP Covers are available with elastic, zippers, snaps and see through windows and Identification Pouches
pp_muff3-removebg.png
Polypropylene muffs are a great choice in non-sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover to protect process hoses.
082.png
Save time and standardize the activities in equipment preparation for sterilization. Leave the covers in place during installation to prevent microbial and particulate contamination.
297.png
PureClean™ Muffs are made with a new untreated, flexible Tyvek® material specifically designed for pharmaceutical use in steam sterilization applications. Uncoated Tyvek® eliminates the potential of residue deposits on equipment, post sterilization.
634.png
Leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed. Uncoated Tyvek® eliminates the potential of residue deposits on equipment, post sterilization.