Orders LinkContactUsLinkComponent
Autoclave-Bags | Avantor
Autoclave Bags
ss 1421 film bag2.png
PureClean™ self-sealing autoclave bags are made from a transparent film and 1421B DuPont™ Tyvek®, which is a new grade of uncoated, untreated, flexible Tyvek® material specifically designed for pharmaceutical use in steam sterilization applications. Uncoated Tyvek® eliminates the potential of residue deposits on equipment, post sterilization.
179.png
Tyvek® 1422A autoclave bags with steam indicators can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
SS_1422a-removebg-preview.png
Self-sealing Tyvek® 1422A autoclave bags with steam indicators can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
1422A Tyvek Drawstring Autoclave Bag.png
The drape-able nature of the Tyvek® 1422A material is ideal for wrapping equipment parts where one may need to 'feel through' the bag; especially when lifting, or handling the wrapped part.
170.png
Continuous Form tubing made from 1422A Tyvek® provides the user with tremendous sizing flexibility when wrapping pharmaceutical equipment for steam sterilization.
467.png
Protect your filling needles from accidental touching after they are removed from the primary sterilization packaging. Also creates secondary level of protection during aseptic presentation.
paper pouch before.png
Designed for light duty, these economical sterilization pouches are designed for steam sterilization cycles (121 to 135 °C)
674.png
These bags protect filling needles from accidental touching after they are removed from the primary sterilization packaging.
091.png
The use of these 2 ply self-sealing bags will save valuable time in the equipment prep area. The 2-ply BHD material resists ripping and punctures.
226 (2).png
Standardize and simplify the way equipment is wrapped for steam sterilization.
easy tear3 (1).jpg.png
Easy-Tear Feature for easier opening and sealing.  No sterile cutting instruments required to open the bag
Self-Sealing 1073B.png
A significant upgrade over paper based steam sterilization pouches. These self-sealing Tyvek® peel pouches will not rip or puncture like paper based autoclave pouches routinely do.
Heat Seal 1073B.png
These heat seal Tyvek® peel pouches will not rip or puncture like paper based autoclave pouches routinely do
Continuous Form 1073B Tyvek.png
Unroll and cut to desired length, heat seal both ends.  A significant upgrade over paper-based steam sterilization reels
Autoclavable at 121 to 125 °C
1073B Tyvek Stopper.png
Stopper bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps. Also a good choice for heavy equipment parts where visibility into the bag is required.
1073B Tyvek Stopper Bag.png
Unroll and cut to desired length, heat seal both ends. Stopper bag tubing is ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps.
All 1073B Tyvek Heat Seal.png
100% Medical Grade 1073B Tyvek® Autoclave Bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps.
Self-Sealing, All 1073B.png
Self-sealing closure reduces prep time and eliminates the need for and the validation of a heat sealing machine.
Self-Sealing Heavy Duty.png
Designed for higher temperature steam sterilization cycles (135 °C)* and compatible with ethylene oxide and hydrogen peroxide sterilization.
N179.png
PureClean™ Autoclave bags are made from 1421B DuPont™ Tyvek®, which is a grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
N_SS_1422a-removebg-preview.png
Self-sealing PureClean™ Autoclave bags are made from 1421B DuPont™ Tyvek®, which is a grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
N_Tyvek Drwastring Bag.png
PureClean™ Autoclave bags with drawstring closures are made from 1421B DuPont™ Tyvek®, which is a grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
N171.png
Continuous Form PureClean™ Tyvek Tubing is made from 1421B DuPont™ Tyvek®, which is a grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process..
N467.png
PureClean™ Tyvek Filling Needle bags are made from 1421B DuPont™ Tyvek®, which is a grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.