Orders LinkContactUsLinkComponent
accuracy1.png
How to improve your weighing accuracy and comply with the changing requirements in highly regulated areas
Webinar
Thursday, October 14, 2021
9:00 am to 10:00 am EST
Replay
How to improve your weighing accuracy and comply with the changing requirements in highly regulated areas | Avantor
Email_Icon_RGB.png
Email alerts
Registration_Icon_RGB.png
Free registration
Chat_Icon_RGB.png
Chat area
Certified_Icon_RGB.png
Certificate of attendance
This webinar gives an overview on best weighing practices and describes factors influencing the weighing result. Balance intrinsic factors such as repeatability, linearity, eccentric load, calibration & adjustment, environmental influences such as climate, leveling, vibrations and influences coming from the operator and samples as magnetism, electric charges and air buoyancy effects are discussed in detail.
Possible effects on the weighing result and preventive measures are presented and there will also be a focus on important news for the pharmaceutical industry including the new Chapter 2.1.7 of the European Pharmacopeia.
Presented by:
Eszter_Paldy_Foto.jpg
Eszter Paldy
Eszter Paldy studied biology and earned her PhD in Pharmacology. During her work at the Pharmacology Institute of the Medical University in Heidelberg in Germany she has further educated herself in business, project – and product management. She has joined Sartorius in 2017. She is a product manager for premium balances and mass comparators and is actively involved in product development projects.
Foto_Densow.jpg
Holger Densow
Holger Densow studied biology at the University of Bielefeld, Germany and finished his Doctorate in 2005. Then he was working as sales representative for 2,5 years for a German Biotech company and in 2008 switched to another company to manage lab instruments as product manager. Since 2016 he is working as Product Manager at Sartorius and he is taking care of Software Solutions for Lab Balances.