Orders LinkContactUsLinkComponent
32363-1920x760.jpg
Microbiological “Gold standards” VS Flow cytometry method in Food & Beverage industry
Webinar
Tuesday, May 25, 2021  
16:00 to 17:00 CEST (Berlin, Paris, Madrid)
Replay
Rapid Enumeration of Microorganisms | Avantor
Email_Icon_RGB.png
Email alerts
Registration_Icon_RGB.png
Free registration
Chat_Icon_RGB.png
Chat area
Certified_Icon_RGB.png
Certificate of attendance
Flow cytometry (FCM) has become a valuable tool in food microbiology. It has been developed to rapidly enumerate microorganisms, to distinguish between viable, metabolically-active and dead cells, which is of great importance in food development and food spoilage.
Currently, the food industry is still using quality control analytical techniques based on plating of diluted samples onto media and counting the resultant colonies after incubation under defined condition of time and temperature. This method is labour and time consuming. In the case of certain pathogens such as E. coli, this process can take up to 5 days. Moreover, further biochemical identification can prolong the duration for up to 7 days.
For more information, please contact us at [email protected]
Presented by
Joanna.jpg
Joanna Śliwa-Dominiak
Joanna Śliwa-Dominiak has a Ph.D. in biological sciences, specialising in microbiology from the University of Szczencinski where she studied bacteriophages found in the digestive system and their presence in the aquatic environment as indicators of faecal pollution. After her PhD she went on to lecture in microbiology and microbial genetics at the University of Szczencinski.
Katarzyna.jpg
Katarzyna Sielatycka
Katarzyna Sielatycka, has Ph.D., from Pomeranian Medical University in Szczecin, where she studied in the field of adult stem cell biology and probiotic bacteria. After completing her studies, she conducted fellowship research at the Stem Cell Institute, University of Louisville, USA. Before returning back to Poland.