Orders LinkContactUsLinkComponent
Freeze-drying.png
Freeze drying and sublimation: The importance of precise vacuum technology
Webinar
Tuesday, 11th October, 2022
11:00 to 12:00 CEST (Berlin, Paris, Madrid)
Replay
Freeze drying and sublimation: The importance of precise vacuum technology | Avantor
WebinarEmailAlerts.jpg
Email alerts
WebinarFreeRegistration.jpg
Free registration
Webinar-chat-area.jpg
Chat area
webinar-Certificate-of-attendance.jpg
Certificate of attendance
The goal of freeze drying is to dry samples as gently as possible. The physical principle of sublimation is used to let ice turn directly into a gaseous state without melting water forming. Vacuum plays a key role here, as sublimation is only possible under exact pressure conditions.
In this webinar, our VACUUBRAND expert explains why precise vacuum is crucial in this application and why too low pressure is counterproductive. You will also receive valuable advice on how to not only achieve optimum results and process times, but also how to effectively protect your vacuum pump from wear.
  • The principle of freeze drying and sublimation explained
  • Why exact vacuum is key
  • Tips for optimal process performance
  • Selection of the correct vacuum technology
  • Tips for protecting the vacuum pump against wear
For more information, please contact us at [email protected]
Presented by
Carsten-Ruth.png
Carsten Ruth
With chemical engineering background and experience in the fields of chemistry, analytics, and material sciences, Carsten Ruth joined VACUUBRAND in 2005. Combining technical background, application knowledge, and experience gained from customer’s input, Carsten could built up an expertise experience level for vacuum technologies and application solutions. With his extensive experience in sales, customer relations, and distribution management Carsten is leading the operative sales department of VACUUBRAND in a global team.