Orders LinkContactUsLinkComponent
NanoCuvetteHero.png
Achieve easy, fast and reliable particle/cell size analysis with NanoCuvette™ S
Webinar
Tuesday, 23rd November, 2021  
15:30 to 16:30 CEST (Berlin, Paris, Madrid)
Replay
Achieve easy, fast and reliable particle/cell size analysis with NanoCuvette | Avantor
WebinarEmailAlerts.jpg
Email alerts
WebinarFreeRegistration.jpg
Free registration
Webinar-chat-area.jpg
Chat area
webinar-Certificate-of-attendance.jpg
Certificate of attendance
In this webinar we will introduce NanoCuvette™ S by using practical cases and hands-on demonstration.
The NanoCuvette™ S uses the latest nanotechnology and software to measure both size and concentraction of particles and cells. Unlike DLS instruments which become increasing unreliable for larger particles, NanoCuvette™ S maintains its high accuracy for micro-particles. Furthermore, it provides both size and concentration in the same measurement making it the ideal candidate for routine lab measurements.
The NanoCuvette™ S improves the laboratory workflow and instantly upgrades existing cuvette-based UV-Vis spectrophotometers to achieve easy, fast and reliable particle/cell size analysis.
For more information, please contact us at [email protected]
Presented by
Emil.png
Emil Højlund-Nielsen
Emil Højlund-Nielsen holds a PhD in Nanotechnology from the Technical University of Denmark and has previously worked as a researcher on award-winning projects. Today, he is the CEO of cphnano, a labtech company digititalizing and upgrading UV-Vis spectrophotometry with solutions available via resellers and partners in more than 20 countries.