Orders LinkContactUsLinkComponent
TYGON Lab (R-3603) Tubing | Avantor
TYGON Lab (R-3603) Tubing
Advantages:
  • Inexpensive tubing for general laboratory applications.
  • Nonaging, nonoxidizing.
  • Clear for easy flow monitoring.
  • Handles virtually all inorganic chemicals.
  • Low gas permeability.
  • Smooth bore.
  • Good for viscous fluids.
  • High dielectric constant.
Limitations:
  • Limited pumping life
  • Potential leaching of plasticizer.
Application Suitability:
Acids:Good
Alkalies:Good
Organic solvents:Not recommended
Pressure:Good
Vacuum:Good
Viscous fluids:Excellent
Sterile fluids:Poor
Physical Characteristics and Composition:
  • Thermoplastic.
  • PVC-based material with plasticizer.
  • Firm (stiff) material.
  • Transparent, clear.
Temperature Range:
-58 to 165°F (-50 to 74°C)
Meets Classifications:
  • FDA 21 CFR 175.300
Gas Permeability:
cc x mm
x 10-10
(cm2 x sec. x cm Hg
 CO2:360
 H2:97
 O2:80
 N2:40
Cleaning/Sterilization:
  • Sterilize with ethylene oxide (ETO) or autoclave.
  • To autoclave: Coil loosely in nonlinting cloth or paper; autoclave at 250°F (121°C), 15 psi for 30 minutes (tubing will appear milky); air dry at max 150°F (66°C) for 2 to 2.5 hours until clear.