Orders LinkContactUsLinkComponent
Standard Pipe Thread Fittings | Avantor
Standard Pipe Thread Fittings
American Standard Taper Pipe Threads (NPT)
Nominal size
(inside
diameter)
Actual ODThreads per inchLength of engagement
(tightened by hand)
Hand tight turnsWrench makeup turnsLength of effective thread
1/160.3125270.160  0.2611
180.405270.1800.260
¼0.540180.200430.401
380.675180.24030.408
½0.840140.32030.534
¾1.050140.34030.546
11.31511.50.4000.682
1.66011.50.4200.707
1.90011.50.4200.724
22.37511.50.436530.756
2.87580.68231.136
33.50080.766631.2000
4.00080.8211.2500
44.50080.8441.3000
55.56380.9371.4063
See below illustrations of (when viewed at 100%).
Masterflex standard pipe thread fittings