Orders LinkContactUsLinkComponent
Pump Types and Definitions | Avantor
Pump Types and Definitions
Bellows.png
Bellows
These pumps move fluid through a reciprocating bellows cavity that is coupled to a driving rod. Pumps are found in the "Metering" and "Positive Displacement" pump sections.
Centrifugal.png
Centrifugal
A rotating vanned disk attached to a drive shaft moves fluid without pulsation as it spins. The outlet can be restricted without damaging the pump.
Diaphragm.png
Diaphragm
Pulsations of one or two flexible diaphragms displace liquid while check valves control the direction of the fluid flow.
columnCard.png
Drum
A motor attached to an impeller is mounted on top of a drum. The impeller pushes fluid up the tube set when submersed.
Flexible-Liner.png
Flexible Liner
The outer surface of an inert liner and inner surface of a rotating body block create a fluid channel used to gently pump fluids without pulsation.
Flexible-Impeller.png
Flexible Impeller
Elastomeric impeller traps fluid between the impeller blades and a molded housing that sweeps fluid through the pump housing.
Gear.png
Gear
Fluid is trapped between the teeth of two or three rotating gears. Gear pumps are good for high system-pressure applications and are often magnetically driven.
columnCard.png
Hand
Manually powered pumps of various designs used for short term or repetitive liquid transfer applications.
Peristaltic-tubing.png
Peristaltic (tubing)
Fluid only contacts the tubing—rollers of a motor-driven pump head push the fluid along the tubing as they rotate. Noncontaminating and easy to clean.
Piston.png
Piston
Rotating pistons of varying stroke lengths pump fluids through check valves. Good for high-pressure applications.
columnCard.png
Progressing Cavity
The cavity between a screw-style rotor and stator force high-viscosity and particulate-filled fluids through the pump.
columnCard.png
Pressure Washer
Provide high-pressure water, ideal for cleaning and sanitizing.
columnCard.png
Rotary Lobe
Lobe-shaped rotors gently fill with and dispense fluids containing soft particles, solids, and highly viscous fluids. Sanitary construction is perfect for use in the food and beverage industries.
columnCard.png
Rotary Vane
Operate like flexible impeller pumps but use an impeller made of a rigid material useful for high-pressure or low-shear applications.
columnCard.png
Syringe
Infusion or withdrawal syringe pumps provide high pressure and high accuracy for applications such as HPLC.
columnCard.png
Submersible/Immersible
Centrifugal pumps in a specialized housing used for emptying tanks or sumps.