Orders LinkContactUsLinkComponent
FDA Compliant Viton Tubing | Avantor
FDA Compliant Viton Tubing
Advantages:
  • Perfect for food and lab applications where FDA compliance is required
  • Good chemical resistance
  • Resistant to corrosives
  • High temperature properties
Limitations:
  • Limited pumping life
Application Suitability:
Acids:Excellent
Alkalies:Excellent
Organic solvents:Variable—test before using
Pressure:Good
Vacuum:Good
Viscous fluids:Good
Sterile fluids:Fair
Physical Characteristics and Composition:
  • Thermal set rubber
  • Viton B (67 percent fluorine)
  • Firm (stiff) material
  • Opaque, black
Temperature Range:
-25 to 400°F (-32 to 205°C)
Meets Classifications:
  • FDA 21 CFR 177.2600
Gas Permeability:
cc x mm
x 10
(cm2 x sec. x cm Hg
CO2:76 to 79
H2:-
O2:13 to 15
N2:4.3
Cleaning/Sterilization:
  • Sterilize with circulating hot air oven at 249°C (480°F) for 16 hours
Viton - Reg TM DuPont Dow Elastomers L.L.C.