Orders LinkContactUsLinkComponent
 Universal Pattern Guide for AC Power Plugs/Sockets | Avantor
Universal Pattern Guide for AC Power Plugs/Sockets
ACpwrplug.png
Electrical plugs and their sockets differ by country in shape, size, and type of connectors. The current Universal Pattern Guide for AC Power Plugs/Sockets are available for FREE download on this Worldwide Socket pdf file — Complete details categorized alphabetically by country or region.
Some of our pump drives include 6-ft (1.8-m) line cord with IEC 320/CEE22 socket and are shipped with country specific plug/cord set; please order the plug type needed to use in your country. For example of plug types see Masterflex Power Cord Assemblies.