Orders LinkContactUsLinkComponent
vwr-plus-flammable-storage-laboratory-freezer-30-cu-ft955x590.jpg
Controlled Temperature Units
Avantor Equipment and Instrument Service (E&IS) offers installation, validation, calibration, preventative maintenance, and service for all controlled temperature unit makes and models.
Constant Temperature Units | Avantor
Our Service offerings include the following:
  • Ovens
  • Incubators and CO2 Incubators
  • Environmental chambers
  • Water Baths
  • Circulators
  • Chillers
  • Ultra Low Temperature Freezers (ULT)
  • Refrigerators
  • Freezers
  • And more...
Installation
Avantor Equipment and Instrument Service (E&IS) provides installation service to ensure that your equipment is set up, powered on and functioning correctly.
Preventative Maintenance and Calibration
We offer customized Preventative Maintenance plans based on the specific needs of your unit. Preventative maintenance plans help identify problems before they occur to reduce downtime and equipment failures. Calibration services ensure that your equipment runs optimally to ensure accuracy and reliability.
Temperature Mapping
Temperature mapping provides data to understand temperature distribution throughout the temperature-controlled unit. The mapping requirements will vary based on the expected use of each unit and process critical parameters. Data is reviewed to provide an understanding of deviations from min and max temperature points, averages, and other statistical data points relevant to your process.
IQ/OQ/PQ Service
Validation studies of your temperature-controlled units is an important component of a robust qualification plan. Validation protocols include Installation Qualification (IQ), Operational Qualification (OQ), and Performance Qualification (PQ). Temperature mapping is a critical component of this process to ensure operational and performance integrity.
Our qualification services will help you stay in compliance with FDA, GMP/GLP, US Pharmacopoeia and ISO standards.