ContactUsLinkComponent
3M™ Polyimide Film Tape 5413, Gold, 304,8 mm x 32,9 m x 0,07 mm, 1 roll/case 1 * 1 Roll
Catalog # MMMA7000048687
Supplier:  3M
New Product
3M™ Polyimide Film Tape 5413, Gold, 304,8 mm x 32,9 m x 0,07 mm, 1 roll/case 1 * 1 Roll
Catalog # MMMA7000048687
Supplier:  3M