1 component: Benzene [CAS:71-43-2] 2.6g/l in Methanol CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a refrigerator at temperatures between 2°C to 8°C packaging: 5*1ml ampoule 1 * 5 Ampoul
Catalog # CMPLRD0810831.5
Supplier: CUSTOM MADE CHEMICALS LAB