ContactUsLinkComponent
Muffle Furnace LT40/11/C550 Tmax1100 with lift door and over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3047
Supplier:  NABERTHERM
New Product
Muffle Furnace LT40/11/C550 Tmax1100 with lift door and over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3047
Supplier:  NABERTHERM