ContactUsLinkComponent
Muffle Furnace LT40/11/C550 Tmax1100 with lift door and protective gas connection, over-temperature limiter and thermocouple hole in furnace door 1 * 1 items
Artikelnr. 455-3043
Supplier:  NABERTHERM
New Product
Muffle Furnace LT40/11/C550 Tmax1100 with lift door and protective gas connection, over-temperature limiter and thermocouple hole in furnace door 1 * 1 items
Artikelnr. 455-3043
Supplier:  NABERTHERM