ContactUsLinkComponent
Muffle Furnace L9/11/B510 Tmax1100 with flap door and protective gas connection, over-temperature limiter and thermocouple hole in f urnace door 1 * 1 items
Catalog # 455-2719
Supplier:  NABERTHERM
New Product
Muffle Furnace L9/11/B510 Tmax1100 with flap door and protective gas connection, over-temperature limiter and thermocouple hole in f urnace door 1 * 1 items
Catalog # 455-2719
Supplier:  NABERTHERM