ContactUsLinkComponent
uvex super f OTG IR-ex infrared overspecs anti-fog on the inside, extremely scratch-resistant and chemical-resistant on the outside, minim - black/red 1 * 1 items
Catalog # UVEX9169173
Supplier:  UVEX
New Product
uvex super f OTG IR-ex infrared overspecs anti-fog on the inside, extremely scratch-resistant and chemical-resistant on the outside, minim - black/red 1 * 1 items
Catalog # UVEX9169173
Supplier:  UVEX