ContactUsLinkComponent
WEDO® Safety Cutter Plus 839, the Safety Cutter Plus 839 sits comfortably in the hand thanks to the smooth-running, long lever for releasing the blade. Nylon housing with rubberized grip zone, metal blade guide. The spring-loaded blade retracts FULL 1 * 1 items
Catalog # WEDO78839
Supplier:  WEDO WERNER DORSCH
New Product
undefined
WEDO® Safety Cutter Plus 839, the Safety Cutter Plus 839 sits comfortably in the hand thanks to the smooth-running, long lever for releasing the blade. Nylon housing with rubberized grip zone, metal blade guide. The spring-loaded blade retracts FULL 1 * 1 items
Catalog # WEDO78839
Supplier:  WEDO WERNER DORSCH