ContactUsLinkComponent
WEDO® Magnifier with LED-light. With this stylish magnifier you see things more clearly. To switch on the extra bright LED-light please raise the upper part of the housing. The size of the plastic lense is approx. 3.5 x 3.8 cm and provides 3x magnifi 1 * 1 items
Catalog # WEDO2717501
Supplier:  WEDO WERNER DORSCH
New Product
undefined
WEDO® Magnifier with LED-light. With this stylish magnifier you see things more clearly. To switch on the extra bright LED-light please raise the upper part of the housing. The size of the plastic lense is approx. 3.5 x 3.8 cm and provides 3x magnifi 1 * 1 items
Catalog # WEDO2717501
Supplier:  WEDO WERNER DORSCH