ContactUsLinkComponent
Cellcyte 1 - complete imaging system including: control unit with software for device control and acquistion, includes pc & 22 inch hd monitor, 10x objective, well plate tray, 3 fluorescence channels, dapi - ex: 390/40 em: 445/33, gfp - em: 482 1 * 1 items
Catalog # ECHLECCC1200
Supplier:  DISCOVER ECHO
New Product
Cellcyte 1 - complete imaging system including: control unit with software for device control and acquistion, includes pc & 22 inch hd monitor, 10x objective, well plate tray, 3 fluorescence channels, dapi - ex: 390/40 em: 445/33, gfp - em: 482 1 * 1 items
Catalog # ECHLECCC1200
Supplier:  DISCOVER ECHO