ContactUsLinkComponent
Porcine IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an HRP-labelled monoclonal detection antibo dy, Porcine IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair kit can be used to qua 1 * 576 Tests
Artikelnr. ANTIA270532-696
Supplier:  ANTIBODIES.COM
New Product
Porcine IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an HRP-labelled monoclonal detection antibo dy, Porcine IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair kit can be used to qua 1 * 576 Tests
Artikelnr. ANTIA270532-696
Supplier:  ANTIBODIES.COM