ContactUsLinkComponent
Human GM-CSF Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant human GM-CSF ELISA standard, and standard reconstitution buffer. This matched antib 1 * 576 Tests
Catalog # ANTIA270475-696
Supplier:  ANTIBODIES.COM
New Product
Human GM-CSF Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibo dy, Streptavidin-HRP, recombinant human GM-CSF ELISA standard, and standard reconstitution buffer. This matched antib 1 * 576 Tests
Catalog # ANTIA270475-696
Supplier:  ANTIBODIES.COM