ContactUsLinkComponent
Human Mannan Binding Lectin/MBL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Mannan Binding Lectin/MBL in serum, plasma, tissue homogenates, cell lysates, cell cultu 1 * 96 Tests
Catalog # ANTIA2504-96
Supplier:  ANTIBODIES.COM
New Product
Human Mannan Binding Lectin/MBL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Mannan Binding Lectin/MBL in serum, plasma, tissue homogenates, cell lysates, cell cultu 1 * 96 Tests
Catalog # ANTIA2504-96
Supplier:  ANTIBODIES.COM