ContactUsLinkComponent
Human IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an AP-labelled monoclonal detection antibody, native human IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair kit can be used to q 1 * 576 Tests
  ANTIA270397-696
New Product
Human IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an AP-labelled monoclonal detection antibody, native human IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair kit can be used to q 1 * 576 Tests
  ANTIA270397-696