ContactUsLinkComponent
Mouse Anti-SARS-CoV-2 Spike RBD (alpha B.1.1.7) IgM ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vi tro quantitative determination of mouse Anti-SARS-CoV-2 Spike RBD (alpha B.1.1.7) IgM in serum, plasma, tissue homoge 1 * 96 Tests
Catalog # ANTIA303455-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Anti-SARS-CoV-2 Spike RBD (alpha B.1.1.7) IgM ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vi tro quantitative determination of mouse Anti-SARS-CoV-2 Spike RBD (alpha B.1.1.7) IgM in serum, plasma, tissue homoge 1 * 96 Tests
Catalog # ANTIA303455-96
Supplier:  ANTIBODIES.COM