ContactUsLinkComponent
Human alpha 2c Adrenergic Receptor / ADRA2C ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed fo r the in vitro quantitative determination of human alpha 2c Adrenergic Receptor / ADRA2C in serum, plasma, and other 1 * 96 Tests
Catalog # ANTIA310607-96
Supplier:  ANTIBODIES.COM
New Product
Human alpha 2c Adrenergic Receptor / ADRA2C ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed fo r the in vitro quantitative determination of human alpha 2c Adrenergic Receptor / ADRA2C in serum, plasma, and other 1 * 96 Tests
Catalog # ANTIA310607-96
Supplier:  ANTIBODIES.COM