ContactUsLinkComponent
Human Anti-SARS-CoV-2 Spike RBD (alpha B.1.1.7) IgA ELISA Kit is an indirect Enzyme-Linked Immunosorbent Assay designed for the in v itro quantitative determination of human Anti-SARS-CoV-2 Spike RBD (alpha B.1.1.7) IgA in serum, plasma, tissue homog 1 * 96 Tests
Catalog # ANTIA319515-96
Supplier:  ANTIBODIES.COM
New Product
Human Anti-SARS-CoV-2 Spike RBD (alpha B.1.1.7) IgA ELISA Kit is an indirect Enzyme-Linked Immunosorbent Assay designed for the in v itro quantitative determination of human Anti-SARS-CoV-2 Spike RBD (alpha B.1.1.7) IgA in serum, plasma, tissue homog 1 * 96 Tests
Catalog # ANTIA319515-96
Supplier:  ANTIBODIES.COM