ContactUsLinkComponent
Human IL-4I1/LAO ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human IL-4I1/LAO in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples. 1 * 96 Tests
Catalog # ANTIA326664-96
Supplier:  ANTIBODIES.COM
New Product
Human IL-4I1/LAO ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human IL-4I1/LAO in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples. 1 * 96 Tests
Catalog # ANTIA326664-96
Supplier:  ANTIBODIES.COM