ContactUsLinkComponent
Human Epstein Barr Virus EA IgG IF Test is an Immunofluorescence Assay designed for the in vitro qualititative determination of huma n EA EBV IgG in serum and plasma. 1 * 160 Tests
Catalog # ANTIA324364-160
Supplier:  ANTIBODIES.COM
New Product
Human Epstein Barr Virus EA IgG IF Test is an Immunofluorescence Assay designed for the in vitro qualititative determination of huma n EA EBV IgG in serum and plasma. 1 * 160 Tests
Catalog # ANTIA324364-160
Supplier:  ANTIBODIES.COM