ContactUsLinkComponent
Human RNA polymerase II CTD repeat YSPTSPS ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human RNA polymerase II CTD repeat YSPTSPS in serum, plasma, and other bi 1 * 96 Tests
Catalog # ANTIA311638-96
Supplier:  ANTIBODIES.COM
New Product
Human RNA polymerase II CTD repeat YSPTSPS ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human RNA polymerase II CTD repeat YSPTSPS in serum, plasma, and other bi 1 * 96 Tests
Catalog # ANTIA311638-96
Supplier:  ANTIBODIES.COM